2,4-Diamino-6-hydroxypyrimidine

Lieferant: Molekula

Synonyme: 6-Aminoisocytosin, 2,4-Diamino-6-pyrimidinol, 2,4-Diaminopyrimidin-6-ol

10931062-100G
MOLE10931062-100GEA 93.1 EUR
MOLE10931062-100G
2,4-Diamino-6-hydroxypyrimidine
2,4-Diamino-6-hydroxypyrimidine
Formel: C₄H₆N₄O
Molecular Weight: 126,12 g/mol
Siedepunkt: 373 °C (1013 hPa)
Schmelzpunkt: 285…286 °C
MDL: MFCD00006098
CAS-Nummer: 56-06-4
EINECS: 200-254-4
Merck Index: 13,03001

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR