2,6-Dichlorphenol-indophenol Natriumsalz Hydrat
Lieferant: Molekula
Synonyme:
DCPIP Hydrat, Tillmans-Reagenz Hydrat, Dichlorphenolindophenol Hydrat, DPIP Hydrat, Dichlorindophenol Hydrat, DCIP Hydrat, 2,6-Dichlor-N-(4-hydroxyphenyl)-1,4-benzochinonimin Natriumsalz Hydrat, Natrium-4-(3,5-dichlor-4-oxocyclohexa-2,5-dienylidenamino)phenoxid Hydrat
10933059
MOLE10933059EA
0
EUR
MOLE10933059
2,6-Dichlorphenol-indophenol Natriumsalz Hydrat
2,6-Dichlorphenol-indophenol Natriumsalz Hydrat
Formel:
C₁₂H₆Cl₂NNaO₂·xH₂O Molecular Weight: 290,08 g/mol (anhy) Schmelzpunkt: > 300 °C |
MDL:
MFCD00150014 CAS-Nummer: 1266615-56-8 EINECS: 210-640-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...