2,3-Dimethoxybenzylalkohol
Lieferant: Molekula
Synonyme:
(2,3-Dimethoxyphenyl)methanol
11004673-25G
MOLE11004673-25GEA
0
EUR
MOLE11004673-25G
2,3-Dimethoxybenzylalkohol
2,3-Dimethoxybenzylalkohol
Formel:
C₉H₁₂O₃ Molecular Weight: 168,19 g/mol Siedepunkt: 258 °C (1013 hPa) Schmelzpunkt: 50 °C |
MDL:
MFCD00004612 CAS-Nummer: 5653-67-8 EINECS: 227-099-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...