Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3) Dihydrat

Lieferant: Molekula

11038928-1G
MOLE11038928-1GEA 71.6 EUR
MOLE11038928-1G
Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3) Dihydrat
Uridin-5'-triphosphorsäure Trinatriumsalz (UTP-Na3) Dihydrat
Formel: C₉H₁₂N₂Na₃O₁₅P₃·2H₂O
Molecular Weight: 586.12 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 116295-90-0
EINECS: 243-347-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR