(S)-(-)-1,1'-Bi-2-naphthol

Lieferant: Molekula

11057305-1G
MOLE11057305-1GEA 0 EUR
MOLE11057305-1G
(S)-(-)-1,1'-Bi-2-naphthol
(S)-(-)-1,1'-Bi-2-naphthol
Formel: C₂₀H₁₄O₂
MDL: MFCD00004068
CAS-Nummer: 18531-99-2
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR