(Ethyl)triphenylphosphoniumiodid
Lieferant: Molekula
Synonyme:
Ethyltriphenylphosphonium iodide
11076392-100G
MOLE11076392-100GEA
0
EUR
MOLE11076392-100G
(Ethyl)triphenylphosphoniumiodid
(Ethyl)triphenylphosphoniumiodid
Formel:
C₂₀H₂₀IP Molecular Weight: 418,26 g/mol Schmelzpunkt: 163…166 °C Dichte: 1 g/cm³ (20 °C) |
MDL:
MFCD00040352 CAS-Nummer: 4736-60-1 EINECS: 225-245-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...