4-Hydroxy-3-nitropyridin

Lieferant: Molekula

Synonyme: 3-Nitro-4-pyridinol

11085105-25G
MOLE11085105-25GEA 0 EUR
MOLE11085105-25G
4-Hydroxy-3-nitropyridin
4-Hydroxy-3-nitropyridine
Formel: C₅H₄N₂O₃
Molecular Weight: 140,10 g/mol
MDL: MFCD00128874
CAS-Nummer: 5435-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR