CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)

Lieferant: Molekula

Synonyme: 3-(Cyclohexylamino)-2-hydroxy-1-propanesulphonic acid

11091556-500G
MOLE11091556-500GEA 0 EUR
MOLE11091556-500G
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)
Formel: C₉H₁₉NO₄S
Molecular Weight: 237,32 g/mol
Schmelzpunkt: 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041778
CAS-Nummer: 73463-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR