Arecoliniumbromid

Lieferant: Molekula

Synonyme: Arecoline hydrobromide

11161105-25G
MOLE11161105-25GEA 281 EUR
MOLE11161105-25G
Arecoliniumbromid
Arecoliniumbromid
Formel: C₈H₁₄BrNO₂
Molecular Weight: 236.11 g/mol
MDL: MFCD00039041
CAS-Nummer: 300-08-3
EINECS: 206-087-3
UN: 1544
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR