L(+)-Lysin Monohydrochlorid
Lieferant: Molekula
Synonyme:
(S)-(+)-Lysin Monohydrochlorid, H-Lys-OH.HCl, (S)-(+)-2,6-Diaminohexansäure Hydrochlorid, (S)-(+)-2,6-Diaminocapronsäure Hydrochlorid, L(+)-Lysin Hydrochlorid, (S)-(+)-2,6-Diaminohexansäure Monohydrochlorid, (S)-(+)-2,6-Diaminocapronsäure Monohydrochlorid, (S)-(+)-Lysin Hydrochlorid
11195192-1KG
MOLE11195192-1KGEA
164
EUR
MOLE11195192-1KG
L(+)-Lysin Monohydrochlorid
L(+)-Lysin Monohydrochlorid
Formel:
H₂N(CH₂)₄CH(NH₂)CO₂H·HCl Molecular Weight: 182,65 g/mol Schmelzpunkt: 263…264 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064564 CAS-Nummer: 657-27-2 EINECS: 211-519-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...