Triethylammoniumdihydrogentrifluorid
Lieferant: Molekula
Synonyme:
Triethylamin Trishydrofluorid, Triethylamin Trihydrofluorid
11203893-25G
MOLE11203893-25GEA
0
EUR
MOLE11203893-25G
Triethylammoniumdihydrogentrifluorid
Triethylammonium dihydrogentrifluoride
Formel:
C₆H₁₈F₃N Molecular Weight: 161,21 g/mol Dichte: 1,094 g/cm³ (20 °C) |
MDL:
MFCD00043294 CAS-Nummer: 73602-61-6 EINECS: 277-550-5 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...