(2-Bromethyl)ammoniumbromid

Lieferant: Molekula
Achtung

Synonyme: 2-Bromethylamin Hydrobromid, 1-Amino-2-bromethan Hydrobromid

11263214-500G
MOLE11263214-500GEA 0 EUR
MOLE11263214-500G
(2-Bromethyl)ammoniumbromid
2-Bromethylamine hydrobromide
Formel: BrCH₂CH₂NH₃Br
Molecular Weight: 204.89 g/mol
Schmelzpunkt: 170…173 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012886
CAS-Nummer: 2576-47-8
EINECS: 219-924-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR