5-Nitrouracil

Lieferant: Molekula

Synonyme: 2,4-Dihydroxy-5-nitropyrimidin, Nitrouracil

11358007-100G
MOLE11358007-100GEA 130 EUR
MOLE11358007-100G
5-Nitrouracil
5-Nitrouracil
Formel: C₄H₃N₃O₄
Molecular Weight: 157,09 g/mol
Schmelzpunkt: 300 °C
Dichte: 1,807 g/cm³ (20 °C)
MDL: MFCD00006021
CAS-Nummer: 611-08-5
EINECS: 210-250-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR