Methyl-m-toluat
Lieferant: Molekula
Synonyme:
m-Toluylsäuremethylester, 3-Toluylsäuremethylester, 3-Tolylsäuremethylester, 3-Methylbenzoesäuremethylester, Methyl-3-methylbenzoat
11372927-100G
MOLE11372927-100GEA
127
EUR
MOLE11372927-100G
Methyl-m-toluat
Methyl-m-toluat
Formel:
C₉H₁₀O₂ Molecular Weight: 150,18 g/mol |
MDL:
MFCD00008436 CAS-Nummer: 99-36-5 EINECS: 202-753-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...