1-(Methylamino)-1-(methylthio)-2-nitroethen

Lieferant: Molekula

Synonyme: 1-Methylthio-1-methylamino-2-nitroethylene, N-Methyl-1-(methylthio)-2-nitroethenamine

11406597-25G
MOLE11406597-25GEA 0 EUR
MOLE11406597-25G
1-(Methylamino)-1-(methylthio)-2-nitroethen
1-(Methylamino)-1-(methylthio)-2-nitroethene
Formel: C₄H₈N₂O₂S
Molecular Weight: 148,18 g/mol
MDL: MFCD00040994
CAS-Nummer: 61832-41-5
EINECS: 263-266-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR