3-Chlor-2-fluorbenzoesäure

Lieferant: Molekula

11416359-1G
MOLE11416359-1GEA 0 EUR
MOLE11416359-1G
3-Chlor-2-fluorbenzoesäure
3-Chloro-2-fluorobenzoic acid
Formel: C₇H₄ClFO₂
MDL: MFCD00042506
CAS-Nummer: 161957-55-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR