N(α)-Benzyloxycarbonyl-L-lysin
Lieferant: Molekula
Synonyme:
(S)-6-Amino-2-(((benzyloxy)carbonyl)amino)hexanoic acid, Z-Lys-OH
11422112-5G
MOLE11422112-5GEA
64.5
EUR
MOLE11422112-5G
N(α)-Benzyloxycarbonyl-L-lysin
N(α)-Benzyloxycarbonyl-L-lysin
Formel:
C₁₄H₂₀N₂O₄ Molecular Weight: 280,3 g/mol |
MDL:
MFCD00038204 CAS-Nummer: 2212-75-1 EINECS: 218-662-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...