2-Chlor-5-nitroanilin
Lieferant: Molekula
|
Gefahr
|
11488702-100G
MOLE11488702-100GEA
0
EUR
MOLE11488702-100G
2-Chlor-5-nitroanilin
2-Chlor-5-nitroanilin
Formel:
C₆H₅ClN₂O₂ Molecular Weight: 172,57 g/mol Schmelzpunkt: 119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007668 CAS-Nummer: 6283-25-6 EINECS: 228-498-7 UN: 2237 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...