N,N-Dimethylbenzamid
Lieferant: Molekula
11521589-100G
MOLE11521589-100GEA
0
EUR
MOLE11521589-100G
N,N-Dimethylbenzamid
N,N-Dimethylbenzamid
Formel:
C₉H₁₁NO Molecular Weight: 149,19 g/mol Siedepunkt: 256 °C (1013 hPa) Schmelzpunkt: 40…42 °C Dichte: 1,11 g/cm³ (23 °C) |
MDL:
MFCD00008320 CAS-Nummer: 611-74-5 EINECS: 210-279-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...