2'-Hydroxy-5'-methylacetanilid

Lieferant: Molekula

11580911-5G
MOLE11580911-5GEA 0 EUR
MOLE11580911-5G
2'-Hydroxy-5'-methylacetanilid
2'-Hydroxy-5'-methylacetanilid
Formel: C₉H₁₁NO₂
MDL: MFCD00020183
CAS-Nummer: 6375-17-3
EINECS: 228-934-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR