(-)-Chinin

Lieferant: Molekula

Synonyme: (R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol

11658195-25G
MOLE11658195-25GEA 112 EUR
MOLE11658195-25G
(-)-Chinin
(-)-Chinin
Formel: C₂₀H₂₄N₂O₂
Molecular Weight: 324,42 g/mol
Schmelzpunkt: 176 °C
MDL: MFCD00198096
CAS-Nummer: 130-95-0
EINECS: 205-003-2
Merck Index: 12,08245

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR