Chromon-3-carbonsäure

Lieferant: Molekula

11667804-1G
MOLE11667804-1GEA 0 EUR
MOLE11667804-1G
Chromon-3-carbonsäure
Chromon-3-carbonsäure
Formel: C₁₀H₆O₄
Molecular Weight: 190,15 g/mol
MDL: MFCD00017338
CAS-Nummer: 39079-62-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR