2-Pyrrolcarbaldehyd
Lieferant: Molekula
Synonyme:
Pyrrol-2-carboxaldehyd, 1H-Pyrrol-2-carbaldehyd, Pyrrol-2-carbaldehyd, 1H-Pyrrol-2-carboxaldehyd
11702992-25G
MOLE11702992-25GEA
0
EUR
MOLE11702992-25G
2-Pyrrolcarbaldehyd
2-Pyrrolcarbaldehyd
Formel:
C₅H₅NO Molecular Weight: 95,1 g/mol Siedepunkt: 217 °C (1013 hPa) Schmelzpunkt: 43…46 °C |
MDL:
MFCD00005217 CAS-Nummer: 1003-29-8 EINECS: 213-705-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...