4,5-Dimethylthiazol
Lieferant: Molekula
11765967-25G
MOLE11765967-25GEA
0
EUR
MOLE11765967-25G
4,5-Dimethylthiazol
4,5-Dimethylthiazole
Formel:
C₅H₇NS Molecular Weight: 113,18 g/mol Siedepunkt: 149…153 °C (1013 hPa) Dichte: 1,0828 g/cm³ (20 °C) |
MDL:
MFCD00005336 CAS-Nummer: 3581-91-7 EINECS: 222-703-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...