3-Acetylpyrrol

Lieferant: Molekula

11767384-1G
MOLE11767384-1GEA 0 EUR
MOLE11767384-1G
3-Acetylpyrrol
3-Acetylpyrrol
Formel: C₆H₇NO
Molecular Weight: 109,13 g/mol
MDL: MFCD00067759
CAS-Nummer: 1072-82-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR