tri-Ammoniumorthophosphat Trihydrat

Lieferant: Molekula

11810524-1KG
MOLE11810524-1KGEA 0 EUR
MOLE11810524-1KG
tri-Ammoniumorthophosphat Trihydrat
tri-Ammoniumorthophosphat Trihydrat
Formel: H₁₂N₃O₄P·3H₂O
CAS-Nummer: 25447-33-0
EINECS: 233-793-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR