Bromcyclopropan

Lieferant: Molekula

Synonyme: Cyclopropylbromid

11909051-25G
MOLE11909051-25GEA 0 EUR
MOLE11909051-25G
Bromcyclopropan
Bromcyclopropan
Formel: C₃H₅Br
Molecular Weight: 120,98 g/mol
Siedepunkt: 67…69 °C (1013 hPa)
Dichte: 1,505 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00001271
CAS-Nummer: 4333-56-6
EINECS: 224-375-7
UN: 1993
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR