AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid)
Lieferant: Molekula
|
Achtung
|
Synonyme:
4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride
11939321-500MG
MOLE11939321-500MGEA
348
EUR
MOLE11939321-500MG
AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid)
AEBSF Hydrochlorid (4-(2-Aminoethyl)benzolsulfonylfluorid Hydrochlorid)
Formel:
C₈H₁₁ClFNO₂S Molecular Weight: 239,69 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00132962 CAS-Nummer: 30827-99-7 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...