Dextran, M.W. ~ 40,000

Lieferant: Molekula

11939843-100G
MOLE11939843-100GEA 276 EUR
MOLE11939843-100G
Dextran, M.W. ~ 40,000
Dextran
Formel: (C₆H₁₀O₅)n
MDL: MFCD00130935
CAS-Nummer: 9004-54-0
EINECS: 232-677-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR