2-Brombenzolsulfonylchlorid

Lieferant: Molekula

Synonyme: 2-Brombenzolsulfochlorid, 2-Brombenzolsulfonsäurechlorid, 2-Brombenzol-1-sulfonylchlorid

12081018-5G
MOLE12081018-5GEA 0 EUR
MOLE12081018-5G
2-Brombenzolsulfonylchlorid
2-Brombenzolsulfonylchlorid
Formel: C₆H₄BrClO₂S
Molecular Weight: 255,52 g/mol
Siedepunkt: 127…128 °C (3,6 mmHg)
Schmelzpunkt: 51 °C
MDL: MFCD00051971
CAS-Nummer: 2905-25-1
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR