L(-)-Tyrosin

Lieferant: Molekula

Synonyme: Tyr, (S)-(-)-2-Amino-3-(4-hydroxyphenyl)propionsäure, L(-)-p-Hydroxyphenylalanin, (S)-(-)-p-Hydroxyphenylalanin, Y, H-Tyr-OH

12097155-1KG
MOLE12097155-1KGEA 176 EUR
MOLE12097155-1KG
L(-)-Tyrosin
L(-)-Tyrosin
Formel: C₉H₁₁NO₃
Molecular Weight: 181.19 g/mol
Siedepunkt: 351
Schmelzpunkt: 285
Dichte: 1.41 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002606
CAS-Nummer: 60-18-4
EINECS: 200-460-4
Merck Index: 13,09907

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR