1-(2-Chlorethyl)piperidiniumchlorid

Lieferant: Molekula

Synonyme: N-(2-Chloroethyl)-piperidine hydrochloride, 1-(2-Chloroethyl)piperidine hydrochloride

12162979-500G
MOLE12162979-500GEA 0 EUR
MOLE12162979-500G
1-(2-Chlorethyl)piperidiniumchlorid
1-(2-Chlorethyl)piperidiniumchlorid
Formel: C₇H₁₅Cl₂N
Molecular Weight: 184,11 g/mol
MDL: MFCD00012837
CAS-Nummer: 2008-75-5
EINECS: 217-920-5
UN: 2928
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR