Benzamidin Hydrochlorid Hydrat

Lieferant: Molekula
Achtung

Synonyme: Phenylamidin Hydrochlorid Hydrat, Benzoesäureamidin Hydrochlorid Hydrat, Benzamidiniumchlorid Hydrat

12268137-100G
MOLE12268137-100GEA 507 EUR
MOLE12268137-100G
Benzamidin Hydrochlorid Hydrat
Benzamidin Hydrochlorid Hydrat
Formel: C₆H₅C(=NH)NH₂·HCl·H₂O
Molecular Weight: 174,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00066285
CAS-Nummer: 206752-36-5
EINECS: 216-795-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR