2'-Hydroxyacetanilid
Lieferant: Molekula
Synonyme:
Ethansäure(2-hydroxyphenyl)amid, N-Acetyl-2-aminophenol, N-(2-Hydroxyphenyl)acetamid, 2-Acetamidophenol
12274085-100G
MOLE12274085-100GEA
180
EUR
MOLE12274085-100G
2'-Hydroxyacetanilid
2'-Hydroxyacetanilid
Formel:
C₈H₉NO₂ Molecular Weight: 151,16 g/mol |
MDL:
MFCD00002181 CAS-Nummer: 614-80-2 EINECS: 210-396-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...