2-(3-Bromphenoxy)tetrahydropyran

Lieferant: Molekula

12290112-25G
MOLE12290112-25GEA 0 EUR
MOLE12290112-25G
2-(3-Bromphenoxy)tetrahydropyran
2-(3-Bromphenoxy)tetrahydropyran
Formel: C₁₁H₁₃BrO₂
MDL: MFCD00094009
CAS-Nummer: 57999-49-2
EINECS: 429-030-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR