Cytidin-5'-triphosphorsäure Dinatriumsalz (CTP-Na2) Dihydrat

Lieferant: Molekula

Synonyme: Sodium ((2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogentriphosphate dihydrate, Cytidine-5'-Triphosphate disodium salt dihydrate

12449118-100MG
MOLE12449118-100MGEA 0 EUR
MOLE12449118-100MG
Cytidin-5'-triphosphorsäure Dinatriumsalz (CTP-Na2) Dihydrat
Cytidin-5'-triphosphorsäure Dinatriumsalz (CTP-Na2) Dihydrat
Formel: C₉H₁₄N₃Na₂O₁₄P₃·2H₂O
Molecular Weight: 527,13 g/mol (anhy)
MDL: MFCD00150807
CAS-Nummer: 81012-87-5
EINECS: 252-849-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR