2,4-Dibromthiophen
Lieferant: Molekula
12518574-1G
MOLE12518574-1GEA
0
EUR
MOLE12518574-1G
2,4-Dibromthiophen
2,4-Dibromothiophene
Formel:
C₄H₂Br₂S Molecular Weight: 241,93 g/mol Siedepunkt: 215 °C (1013 hPa) Schmelzpunkt: –30 °C Dichte: 2,14 g/cm³ (20 °C) |
MDL:
MFCD00043889 CAS-Nummer: 3140-92-9 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...