6-Nitro-1-indanon

Lieferant: Molekula

Synonyme: 6-Nitro-2,3-dihydro-1H-inden-1-one

12551217-1G
MOLE12551217-1GEA 180 EUR
MOLE12551217-1G
6-Nitro-1-indanon
6-Nitro-1-indanon
Formel: C₉H₇NO₃
Molecular Weight: 177,16 g/mol
MDL: MFCD06656903
CAS-Nummer: 24623-24-3
EINECS: 246-366-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR