2,4-Dihydroxy-3,6-dimethylbenzoesäure

Lieferant: Molekula

12592063-25G
MOLE12592063-25GEA 0 EUR
MOLE12592063-25G
2,4-Dihydroxy-3,6-dimethylbenzoesäure
2,4-Dihydroxy-3,6-dimethylbenzoic acid
Formel: C₉H₁₀O₄
Molecular Weight: 182,18 g/mol
Dichte: 1 g/cm³ (20 °C)
MDL: MFCD00239340
CAS-Nummer: 4707-46-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR