Benzo[b]thiophen

Lieferant: Molekula

Synonyme: Thianaphthene, 1-Benzothiophene

12658390-100G
MOLE12658390-100GEA 328 EUR
MOLE12658390-100G
Benzo[b]thiophen
Benzo[b]thiophen
Formel: C₈H₆S
Molecular Weight: 134,2 g/mol
MDL: MFCD00005864
CAS-Nummer: 95-15-8
EINECS: 202-395-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR