Kalium-tert-butoxid
Lieferant: Molekula
|
Gefahr
|
Synonyme:
Kalium-tert-butanolat, 2-Methyl-2-propanol Kaliumsalz, Kalium-tert-butylat
12739396-500G
MOLE12739396-500GEA
242
EUR
MOLE12739396-500G
Kalium-tert-butoxid
Kalium-tert-butanolat
Formel:
C₄H₉KO Molecular Weight: 112.21 g/mol Schmelzpunkt: 256…258 °C |
MDL:
MFCD00012162 CAS-Nummer: 865-47-4 EINECS: 212-740-3 UN: 3206 ADR: 4.2 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...