7-Ethylindol

Lieferant: Molekula

Synonyme: 7-Ethyl-1H-indole

12743252-25G
MOLE12743252-25GEA 0 EUR
MOLE12743252-25G
7-Ethylindol
7-Ethylindol
Formel: C₁₀H₁₁N
Molecular Weight: 145,2 g/mol
Siedepunkt: 230 °C (1013 hPa)
Dichte: 1,05 g/cm³ (20 °C)
MDL: MFCD00143514
CAS-Nummer: 22867-74-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR