Dichlor(phenyl)silan

Lieferant: Molekula

12798988-25ML
MOLE12798988-25MLEA 0 EUR
MOLE12798988-25ML
Dichlor(phenyl)silan
Formel: C₆H₆Cl₂Si
MDL: MFCD00039296
CAS-Nummer: 1631-84-1
EINECS: 216-635-3
UN: 2986
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR