Prednisolon
Lieferant: Molekula
Synonyme:
Prednisolonum, 1,2-Dehydrocortisol, 1,4-Pregnadien-11β,17α,21-triol-3,20-dion
12896812-5G
MOLE12896812-5GEA
256
EUR
MOLE12896812-5G
Prednisolon
Prednisolon
Formel:
C₂₁H₂₈O₅ Molecular Weight: 360,45 g/mol Siedepunkt: 571 °C (1013 hPa) Schmelzpunkt: 246 °C Dichte: 1,212 g/cm³ (20 °C) |
MDL:
MFCD00003649 CAS-Nummer: 50-24-8 EINECS: 200-021-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...