Thymolblau
Lieferant: Molekula
Synonyme:
4-[3-(4-Hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxobenzo[c]oxathiol-3-yl]-5-methyl-2-propan-2-ylphenol, Thymolsulfonphthalein
12900344-25G
MOLE12900344-25GEA
71.6
EUR
MOLE12900344-25G
Thymolblau
Thymolblau
Formel:
C₂₇H₃₀O₅S Molecular Weight: 466,6 g/mol Schmelzpunkt: 221 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005869 CAS-Nummer: 76-61-9 EINECS: 200-973-3 Merck Index: 12,09541 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...