5-Benzyloxyindol
Lieferant: Molekula
Synonyme:
5-Benzyloxy-1H-indol
12928137-5G
MOLE12928137-5GEA
68.2
EUR
MOLE12928137-5G
5-Benzyloxyindol
5-Benzyloxyindol
Formel:
C₁₅H₁₃NO Molecular Weight: 223,27 g/mol Siedepunkt: 165 °C (0,1 mmHg) Schmelzpunkt: 107 °C |
MDL:
MFCD00005676 CAS-Nummer: 1215-59-4 EINECS: 214-930-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...