Iminostilben

Lieferant: Molekula

Synonyme: 5H-Dibenzo[b,f]azepine

12960416-100G
MOLE12960416-100GEA 0 EUR
MOLE12960416-100G
Iminostilben
Iminostilben
Formel: C₁₄H₁₁N
Molecular Weight: 193,25 g/mol
MDL: MFCD00005071
CAS-Nummer: 256-96-2
EINECS: 205-970-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR