D(+)-Glucose

Lieferant: Molekula

Synonyme: Dextrose, Traubenzucker

13002238
MOLE13002238EA 0 EUR
MOLE13002238
D(+)-Glucose
D(+)-Glucose
Formel: C₆H₁₂O₆
Molecular Weight: 180.16 g/mol
Schmelzpunkt: 146
Dichte: 1.54 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00148912
CAS-Nummer: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR