Biphenyl-4,4'-dicarbonsäure
Lieferant: Molekula
Synonyme:
4,4'-Biphenyldicarboxylic acid, [1,1'-Biphenyl]-4,4'-dicarboxylic acid
13329950-5G
MOLE13329950-5GEA
0
EUR
MOLE13329950-5G
Biphenyl-4,4'-dicarbonsäure
Biphenyl-4,4'-dicarbonsäure
Formel:
C₁₄H₁₀O₄ Molecular Weight: 242,23 g/mol |
MDL:
MFCD00002554 CAS-Nummer: 787-70-2 EINECS: 212-328-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...