Phenolphthalein
Lieferant: Molekula
|
Gefahr
|
Synonyme:
3,3-Bis(4-hydroxyphenyl)phthalid, 3,3-Bis(4-hydroxyphenyl)-1(3H)-isobenzofuranon
13440009-1KG
MOLE13440009-1KGEA
256
EUR
MOLE13440009-1KG
Phenolphthalein
Phenolphthalein
Formel:
C₂₀H₁₄O₄ Molecular Weight: 318.33 g/mol Schmelzpunkt: 258…262 °C Dichte: 1.3 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005913 CAS-Nummer: 77-09-8 EINECS: 201-004-7 REACH: 01-2119498295-24 Merck Index: 14,07243 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...